Skip to Content
Merck
All Photos(1)

Key Documents

291633

Sigma-Aldrich

HY-STOR® 301 magnesium-nickel alloy

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
CZK 3,780.00

CZK 3,780.00


Please contact Customer Service for Availability


Select a Size

Change View
100 G
CZK 3,780.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.24

CZK 3,780.00


Please contact Customer Service for Availability

type

for hydrogen storage

SMILES string

[Mg].[Ni]

InChI

1S/Mg.Ni

InChI key

ATTFYOXEMHAYAX-UHFFFAOYSA-N

Legal Information

HY-STOR is a registered trademark of Ergenics, Inc.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 1B - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service