291307
4-Iodobutyl acetate
technical grade, 85%
Synonym(s):
1-Acetoxy-4-iodobutane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CO2(CH2)4I
CAS Number:
Molecular Weight:
242.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade
technical grade
Assay
85%
contains
copper as stabilizer
refractive index
n20/D 1.497 (lit.)
bp
94-95 °C/5.5 mmHg (lit.)
density
1.61 g/mL at 25 °C (lit.)
SMILES string
CC(=O)OCCCCI
InChI
1S/C6H11IO2/c1-6(8)9-5-3-2-4-7/h2-5H2,1H3
InChI key
FLPVVTDEKLGZDZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
4-Iodobutyl acetate has been used in the preparation of:
- tricyclohexyl(acetoxybutyl))phosphonium iodide
- tri-n-hexyl(acetoxybutyl))phosphonium iodide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
195.8 °F - closed cup
Flash Point(C)
91 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Phosphonium ionic liquids: design, synthesis and evaluation of biodegradability.
Atefi F, et al.
Green Chemistry, 11(10), 1595-1604 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service