Skip to Content
Merck
All Photos(1)

Key Documents

289043

Sigma-Aldrich

1,3-Dichloro-5-nitrobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NO2
CAS Number:
Molecular Weight:
192.00
Beilstein:
2208877
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

64-65 °C (lit.)

functional group

chloro
nitro

SMILES string

[O-][N+](=O)c1cc(Cl)cc(Cl)c1

InChI

1S/C6H3Cl2NO2/c7-4-1-5(8)3-6(2-4)9(10)11/h1-3H

InChI key

RNABGKOKSBUFHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Dichloro-5-nitrobenzene was tested for fungitoxicity against Aspergillus niger, A. oryzae, Trichoderma viride, Myrothecium verrucaria and Trichophyton mentagrophytes.

Application

1,3-Dichloro-5-nitrobenzene was used as an internal standard in the 1H nuclear magnetic resonance spectroscopic method for the assay of phenylbutazone and oxyphenbutazone.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Husain et al.
Journal - Association of Official Analytical Chemists, 71(3), 525-527 (1988-05-01)
A simple, specific, and rapid 1H nuclear magnetic resonance spectroscopic method for the assay of phenylbutazone and oxyphenbutazone is described. Spectra are recorded in CDCI3 containing 1,3-dichloro-5-nitrobenzene as an internal standard. The aromatic proton resonances for the standard, at delta
H Gershon et al.
Applied microbiology, 22(3), 438-440 (1971-09-01)
Series of 1,3-dihalogeno-5-nitrobenzenes, 3- and 3,5-halogenoanilines, and 2,6-dihalogeno-4-nitroanilines were tested for fungitoxicity against Aspergillus niger, A. oryzae, Trichoderma viride, Myrothecium verrucaria, and Trichophyton mentagrophytes in shaken culture by using Sabouraud dextrose broth enriched with yeast extract as the test medium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service