Skip to Content
Merck
All Photos(1)

Key Documents

282553

Sigma-Aldrich

(1R,2S)-(−)-Norephedrine

99%

Synonym(s):

(1R,2S)-2-Amino-1-phenyl-1-propanol, L-(−)-Norephedrine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CH(CH3)NH2
CAS Number:
Molecular Weight:
151.21
Beilstein:
2207678
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

99%

form

solid

optical activity

[α]20/D −41°, c = 7 in 1 M HCl

mp

51-53 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@H](N)[C@H](O)c1ccccc1

InChI

1S/C9H13NO/c1-7(10)9(11)8-5-3-2-4-6-8/h2-7,9,11H,10H2,1H3/t7-,9-/m0/s1

InChI key

DLNKOYKMWOXYQA-CBAPKCEASA-N

Gene Information

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John S Chappell et al.
Forensic science international, 195(1-3), 108-120 (2009-12-29)
A primary concern with the forensic analysis of the khat plant (Catha edulis) has been the need to preserve the principle psychoactive component, cathinone, which converts to the less-active substance, cathine, after harvesting. The loss of cathinone has serious legal
Hyeon-Kyu Lee et al.
The Journal of organic chemistry, 77(12), 5454-5460 (2012-05-25)
Each of the enantiomers of both norephedrine and norpseudoephedrine were stereoselectively prepared from the common, prochiral cyclic sulfamidate imine of racemic 1-hydroxy-1-phenyl-propan-2-one by employing asymmetric transfer hydrogenation (ATH) catalyzed by the well-defined chiral Rh-complexes, (S,S)- or (R,R)-Cp*RhCl(TsDPEN), and HCO(2)H/Et(3)N as
Susan A Adeoya-Osiguwa et al.
Human reproduction (Oxford, England), 20(1), 198-207 (2004-10-30)
Cathinone, released when Catha edulis leaves (khat) are chewed, has euphoric, stimulatory properties. It is metabolized to the phenylpropanolamines (PPAs) cathine and norephedrine. This study investigated whether PPAs affect mammalian sperm function, using primarily mouse, but also human, spermatozoa. Uncapacitated
Saba Kassim et al.
Journal of ethnopharmacology, 140(1), 193-196 (2012-01-17)
Khat chewing amongst the UK communities originating from Yemen and the East African coast is suggested to create dependency through its main stimulant components (cathinone, norephedrine and norpseudoephedrine) on the central nervous system. To validate self-reported khat chewing behaviours by
A Pokrywka et al.
International journal of sports medicine, 30(8), 569-572 (2009-04-22)
Pseudoephedrine (PSE) as a sympathomimetic is an ingredient of many proprietary medicines which are available on the medical market over the counter (OTC drugs). It can be converted to cathine (CATH, norpseudoephedrine) inside the body. Until the end of 2003

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service