Skip to Content
Merck
All Photos(1)

Documents

280143

Sigma-Aldrich

2-Amino-α-(methoxyimino)-4-thiazoleacetic acid, predominantly syn

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N3O3S
CAS Number:
Molecular Weight:
201.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

192 °C (dec.) (lit.)

SMILES string

CO\N=C(\C(O)=O)c1csc(N)n1

InChI

1S/C6H7N3O3S/c1-12-9-4(5(10)11)3-2-13-6(7)8-3/h2H,1H3,(H2,7,8)(H,10,11)/b9-4+

InChI key

NLARCUDOUOQRPB-RUDMXATFSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total Synthesis of Sodium (3R, 4S)-3-(2-(2-Aminothiazol-4-yl)-(Z)-2-methoxyiminoacetamido)-4-methoxymethyl-2-azetidinone-1-sulfonate from L-Aspartic Acid.
Chung BY, et al.
Bull. Korean Chem. Soc., 13(3), 311-314 (1992)
Total Synthesis of Sodium (3S, 4R)-3-[2-(2-Aminothiazol-4-yl)-(Z)-2-methoxyiminoacetamido]-4-methoxymethyl-2-azetidinone-1-sulfonate from D-Aspartic Acid.
Chung BY, et al.
Bull. Korean Chem. Soc., 13(3), 315-316 (1992)
J V Uri et al.
Acta microbiologica Hungarica, 39(3-4), 317-322 (1992-01-01)
Diazald, a chemical intermediate for the synthesis of biologically active compounds, was found to be a potent in vitro antimicrobial agent against yeasts, yeast-like and filamentous fungi as well as Gram-positive and Gram-negative bacterial strains. Its activity is not inhibited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service