Skip to Content
Merck
All Photos(1)

Documents

277606

Sigma-Aldrich

Tetrabutylammonium chlorochromate

98%

Synonym(s):

TBACC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(ClCrO3)
CAS Number:
Molecular Weight:
377.91
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

98%

reaction suitability

reagent type: oxidant

mp

181-185 °C (lit.)

SMILES string

[O-][Cr](Cl)(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.ClH.Cr.3O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;;;;;/h5-16H2,1-4H3;1H;;;;/q+1;;+1;;;-1/p-1

InChI key

BRRDJFRNQNZSFH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service