271578
N,N-Dimethyl-p-phenylenediamine monohydrochloride
99%
Synonym(s):
4-(Dimethylamino)aniline monohydrochloride, 4-Amino-N,N-dimethylaniline monohydrochloride, DMPPDA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)2NC6H4NH2·HCl
CAS Number:
Molecular Weight:
172.66
Beilstein:
4546912
EC Number:
MDL number:
UNSPSC Code:
12352103
Recommended Products
Assay
99%
mp
215 °C (dec.) (lit.)
SMILES string
Cl[H].CN(C)c1ccc(N)cc1
InChI
1S/C8H12N2.ClH/c1-10(2)8-5-3-7(9)4-6-8;/h3-6H,9H2,1-2H3;1H
InChI key
KTWNIUBGGFBRKH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service