Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C4H3N3O4
CAS Number:
Molecular Weight:
157.08
UNSPSC Code:
12162002
PubChem Substance ID:
MDL number:
Quality Segment
assay
98%
mp
200 °C (dec.) (lit.)
SMILES string
OC(=O)c1nn[nH]c1C(O)=O
InChI
1S/C4H3N3O4/c8-3(9)1-2(4(10)11)6-7-5-1/h(H,8,9)(H,10,11)(H,5,6,7)
InChI key
TZFOEYRGARRRGO-UHFFFAOYSA-N
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
