Skip to Content
Merck
All Photos(2)

Key Documents

269484

Sigma-Aldrich

Devarda′s alloy

filings, +100 mesh

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
CZK 2,270.00
500 G
CZK 6,390.00

CZK 2,270.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
CZK 2,270.00
500 G
CZK 6,390.00

About This Item

Linear Formula:
AlCuZn
CAS Number:
MDL number:
UNSPSC Code:
11101711
PubChem Substance ID:
NACRES:
NA.23

CZK 2,270.00


Please contact Customer Service for Availability

Request a Bulk Order

form

filings

Quality Level

composition

aluminum, ~45%
copper, ~50%
zinc, ~5%

particle size

+100 mesh

mp

550 °C (lit.)

SMILES string

[Al].[Cu].[Zn]

InChI

1S/Al.Cu.Zn

InChI key

MUBKMWFYVHYZAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service