Skip to Content
Merck
All Photos(1)

Documents

269131

Sigma-Aldrich

2-(2-Bromoethyl)-1,3-dioxane

98%

Synonym(s):

3-Bromopropionaldehyde trimethylene acetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11BrO2
CAS Number:
Molecular Weight:
195.05
Beilstein:
1421628
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.481 (lit.)

bp

67-70 °C/2.8 mmHg (lit.)

solubility

ethanol: soluble 50 (μg/mL)

density

1.431 g/mL at 25 °C (lit.)

SMILES string

BrCCC1OCCCO1

InChI

1S/C6H11BrO2/c7-3-2-6-8-4-1-5-9-6/h6H,1-5H2

InChI key

WMDHQEHPOVOEOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The reaction of Grignard reagent prepared from 2-(2-bromoethyl)-1,3-dioxane and N-tert-butanesulfinyl aldimines was studied.

Application

2-(2-Bromoethyl)-1,3-dioxane was used in the synthesis of ketone adducts 2-[3-oxo-4(S)-(triphenylmethyl) amino-6-methylheptyl]-1,3-dioxane and 2-[3-oxo-4(S)-(triphenylmethyl)amino-6-methylheptyl]-1,3-dioxolane.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R L Johnson et al.
International journal of peptide and protein research, 23(6), 581-590 (1984-06-01)
The Grignard reagents of 2-(2-bromoethyl)-1,3-dioxane and 2-(2-bromoethyl)-1,3-dioxolane readily reacted with the 2-thiopyridyl ester of N-triphenylmethyl-L-leucine to give the ketone adducts 2-[3-oxo-4(S)-(triphenylmethyl) amino-6-methylheptyl]-1,3-dioxane (8a) and 2-[3-oxo-4(S)-(triphenylmethyl) amino-6-methylheptyl]-1,3-dioxolane (8b) in near quantitative yield. When 1 equiv. of the Grignard reagent of 2-(2-bromoethyl)-1,3
Kristin M Brinner et al.
Organic & biomolecular chemistry, 3(11), 2109-2113 (2005-05-27)
A new method for the asymmetric synthesis of 2-substituted pyrrolidines in three steps from commercially available starting materials is described. Addition of the Grignard reagent prepared from 2-(2-bromoethyl)-1,3-dioxane to N-tert-butanesulfinyl aldimines proceeds in high yields and with good diastereoselectivities. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service