Skip to Content
Merck
All Photos(1)

Documents

264253

Sigma-Aldrich

2′,4′-Difluoroacetophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3COCH3
CAS Number:
Molecular Weight:
156.13
Beilstein:
1940741
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.488 (lit.)

bp

80-81 °C/25 mmHg (lit.)

density

1.234 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1ccc(F)cc1F

InChI

1S/C8H6F2O/c1-5(11)7-3-2-6(9)4-8(7)10/h2-4H,1H3

InChI key

QEWHNJPLPZOEKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vibrational spectral analysis of 2′,4′-difluoroacetophenone has been carried out by FT-IR and FT-Raman spectroscopy.

Application

2′,4′-Difluoroacetophenone has been used as starting reagent in the synthesis of difluorinated chalcones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Parimala et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 110, 269-284 (2013-04-13)
In this work, the vibrational spectral analysis was carried out by FT-IR and FT-Raman spectroscopy in the regions of 4000-400 cm(-1) and 3500-100 cm(-1), respectively for 2',4'-difluoroacetophenone (DFAP). The molecular structure, fundamental vibrational frequencies and intensity of the vibrational bands

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service