Skip to Content
Merck
All Photos(2)

Documents

263729

Sigma-Aldrich

2-Amino-2-thiazoline hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N2S · HCl
CAS Number:
Molecular Weight:
138.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

200-202 °C (lit.)

SMILES string

Cl[H].NC1=NCCS1

InChI

1S/C3H6N2S.ClH/c4-3-5-1-2-6-3;/h1-2H2,(H2,4,5);1H

InChI key

CODJLHDDIDUKMD-UHFFFAOYSA-N

General description

Metal complexing properties of 2-amino-2-thiazoline hydrochloride with H+, Mg2+, Ca2+, Ni2+, Mn2+, Cu2+ and Zn2+ has been investigated. 2-Amino-2-thiazoline hydrochloride is an anti-cancer molecule and charge distribution in it has been calculated with a quantum mechanic method.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metal complexing ability of two inducers of reverse transformation.
Huang Z-X, et al.
Inorgorganica Chimica Acta, 56, 41-44 (1981)
Y G Smeyers et al.
Revista espanola de oncologia, 30(4), 515-518 (1983-01-01)
The charge distribution of anti-cancer molecules 4-thiazolidine-carboxylic acid and 2-amino-2-thiazoline hydrochloride was calculated with a CNDO/2 semiempiral quantum mechanic method. The activity seems to be related with the formation of Zn2+ and Mn2+ ions. Both molecules show local isosterism, origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service