Skip to Content
Merck
All Photos(3)

Documents

259063

Sigma-Aldrich

(Methoxycarbonylmethyl)triphenylphosphonium bromide

98%

Synonym(s):

(Carbomethoxymethyl)triphenylphosphonium bromide, NSC 136109

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCOCH2P(Br)(C6H5)3
CAS Number:
Molecular Weight:
415.26
Beilstein:
3812975
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

reaction type: C-C Bond Formation

mp

165-170 °C (dec.) (lit.)

SMILES string

[Br-].COC(=O)C[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C21H20O2P.BrH/c1-23-21(22)17-24(18-11-5-2-6-12-18,19-13-7-3-8-14-19)20-15-9-4-10-16-20;/h2-16H,17H2,1H3;1H/q+1;/p-1

InChI key

VCWBQLMDSMSVRL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for synthesis of:
  • GSK-3 inhibitors using theWittig reaction
  • Substituted furanones for antifungal and cytostatic activities
  • Photochromic dithienylethene derivatives
  • Gem-diiodoalkanes containing allylic alcohols
  • Highly substituted cyclopentenes and brazilanes via Au-catalyzed deoxygenative Nazarov cyclization

Reactant for:
  • Asymmetric hydrogenation of α,β-unsaturated ester-phosphonates
  • Studies of thermal decomposition of phosphonium alkyl ester salts

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service