Skip to Content
Merck
All Photos(3)

Key Documents

249246

Sigma-Aldrich

(−)-Menthyl chloride

98%

Synonym(s):

(1R)-(−)-Menthyl chloride, (1S,2R,4R)-2-Chloro-1-isopropyl-1-methylcyclohexane, [1S-(1α,2β,4β)]-2-Chloro-4-methyl-1-(methylethyl)cyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19Cl
CAS Number:
Molecular Weight:
174.71
Beilstein:
1902297
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

optical activity

[α]20/D −52.4°, neat

refractive index

n20/D 1.4634 (lit.)

bp

101-101.5 °C/21 mmHg (lit.)

mp

−20-−16.5 °C (lit.)

density

0.936 g/mL at 25 °C (lit.)

SMILES string

CC(C)[C@@H]1CC[C@@H](C)C[C@H]1Cl

InChI

1S/C10H19Cl/c1-7(2)9-5-4-8(3)6-10(9)11/h7-10H,4-6H2,1-3H3/t8-,9+,10-/m1/s1

InChI key

OMLOJNNKKPNVKN-KXUCPTDWSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service