Skip to Content
Merck
All Photos(1)

Key Documents

243175

Sigma-Aldrich

(R,R)-(−)-2,4-Pentanediol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2CH(OH)CH3
CAS Number:
Molecular Weight:
104.15
Beilstein:
4290613
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]21/D −40.4°, c = 10 in chloroform

optical purity

ee: 97% (GLC)

bp

111-113 °C/19 mmHg (lit.)

mp

48-50 °C (lit.)

SMILES string

C[C@@H](O)C[C@@H](C)O

InChI

1S/C5H12O2/c1-4(6)3-5(2)7/h4-7H,3H2,1-2H3/t4-,5-/m1/s1

InChI key

GTCCGKPBSJZVRZ-RFZPGFLSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acetalization reagent for ketones[1] and β-keto esters.[2] Used in the synthesis of optically active polyesters.[3] C2 symmetric chiral diol with versatile applications as a chiral auxiliary,[4][5] building block,[6][7][8] and chiral ligand.[9][10]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 57, 5438-5438 (1992)
Polymer Journal, 25, 741-741 (1993)
Tetrahedron Letters, 33, 247-247 (1992)
Stolle, A. et al.
Tetrahedron Letters, 35, 3521-3521 (1994)
Marshall, J.A. Xie, S.
The Journal of Organic Chemistry, 60, 7230-7230 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service