Skip to Content
Merck
All Photos(1)

Key Documents

232920

Sigma-Aldrich

4-Nitrosodiphenylamine

75%, technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHC6H4NO
CAS Number:
Molecular Weight:
198.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

Assay

75%

contains

water as balance

mp

144 °C (dec.) (lit.)

SMILES string

O=Nc1ccc(Nc2ccccc2)cc1

InChI

1S/C12H10N2O/c15-14-12-8-6-11(7-9-12)13-10-4-2-1-3-5-10/h1-9,13H

InChI key

OIJHFHYPXWSVPF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

para-Nitrosodiphenylamine.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 27, 227-233 (1982-04-01)
Investigation of a novel liver alcohol dehydrogenase catalyzed redox-elimination reaction involving arylnitroso substrate analogues.
S C Koerber et al.
Biochemistry, 19(4), 731-738 (1980-02-19)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service