Skip to Content
Merck
All Photos(1)

Key Documents

229229

Sigma-Aldrich

Tetraethyltin

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sn(C2H5)4
CAS Number:
Molecular Weight:
234.95
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.473 (lit.)

bp

181 °C (lit.)

mp

−112 °C (lit.)

density

1.187 g/mL at 25 °C (lit.)

SMILES string

CC[Sn](CC)(CC)CC

InChI

1S/4C2H5.Sn/c4*1-2;/h4*1H2,2H3;

InChI key

RWWNQEOPUOCKGR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Experimental study of sensitizing effects of alkyl compounds of tellurium and tin].
E B Romanova
Gigiena i sanitariia, (5)(5), 48-50 (1993-05-01)
G T Pryor et al.
Neurobehavioral toxicology and teratology, 5(1), 91-117 (1983-01-01)
Single-dose LD10S and LD50S were determined in male, Fischer-344 rats for acrylamide monomer, arsenic trioxide, chlordecone, lead acetate, methylmercury hydroxide, monosodium salicylate, tetraethyl tin, and triethyl lead chloride. Proportions of the single-dose LD10S were used in a subacute study to
G V Zakharian
Ukrains'kyi biokhimichnyi zhurnal (1999 ), 84(4), 70-73 (2012-09-06)
The study of the content of cerebrosides, gangliosides and their hydrolytic degradation product--sphingosine in the rat brain with experimental edema was carried out. In parallel, the cytokine profile of pro- and anti-inflammatory cytokines in the blood of rats with experimental
Alessandra Pagliarani et al.
Toxicology in vitro : an international journal published in association with BIBRA, 20(7), 1145-1153 (2006-04-04)
Organotin effects on the Na-dependent ATPases involved in ionic regulation of aquatic animals are poorly known, in spite of the largely documented contamination of seafood, especially bivalve molluscs. This study deals with the in vitro effect of TBT on the
A E Chester et al.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 187(1), 62-68 (1988-01-01)
Carbon disulfide (CS2), tetraethyl lead (TEL), tetraethyl tin (TeET), dithiothreitol (DTT), and gossypol acetic acid (GAA) significantly decreased brain norepinephrine (NE) in rats. The central dopamine (DA) increased after ip administration of CS2, TEL, and DTT, but decreased after TeET

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service