Skip to Content
Merck
All Photos(1)

Key Documents

227293

Sigma-Aldrich

Cyclohexanecarboxamide

97%

Synonym(s):

Hexahydrobenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CONH2
CAS Number:
Molecular Weight:
127.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

186-188 °C (lit.)

functional group

amide

SMILES string

NC(=O)C1CCCCC1

InChI

1S/C7H13NO/c8-7(9)6-4-2-1-3-5-6/h6H,1-5H2,(H2,8,9)

InChI key

PNZXMIKHJXIPEK-UHFFFAOYSA-N

General description

The enthalpy of dilution of cyclohexanecarboxamide in N,N-dimethylformamide was studied to evaluate the enthalpic interaction coefficient.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dai Han et al.
Journal of integrative neuroscience, 18(3), 245-251 (2019-10-12)
The cognitive impairment associated with schizophrenia is highly prevalent and affects the overall functioning of subjects. The stimulation of the serotonin 1A receptor is a primary characteristic of some atypical antipsychotic drugs. We measured the levels of cognitive impairment using
Qing Zhu et al.
Scientific reports, 6, 19266-19266 (2016-01-15)
Vitexin, a C-glycosylated flavone present in several medicinal herbs, has showed various pharmacological activities including antinociception. The present study investigated the antinociceptive effects of vitexin in a mouse model of postoperative pain. This model was prepared by making a surgical
Enthalpies of dilution of benzamide, cyclohexanecarboxamide, and N-acetyl-N′-methyl-l-cyclohexylalaninamide dissolved in N,N-dimethylformamide at 298.15 K.
The Journal of Chemical Thermodynamics, 21(12), 1301-1306 (1989)
Svetlana I Sofronova et al.
The Journal of endocrinology, 235(2), 137-151 (2017-08-11)
The mechanisms of vascular alterations resulting from early thyroid hormones deficiency are poorly understood. We tested the hypothesis that antenatal/early postnatal hypothyroidism would alter the activity of endothelial NO pathway and Rho-kinase pathway, which are specific for developing vasculature. Dams
Alain Lescoat et al.
Frontiers in immunology, 11, 219-219 (2020-03-07)
Inhalation of crystalline silica (SiO2) is a risk factor of systemic autoimmune diseases such as systemic sclerosis (SSc) and fibrotic pulmonary disorders such as silicosis. A defect of apoptotic cell clearance (i.e., efferocytosis, a key process in the resolution of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service