Skip to Content
Merck
All Photos(1)

Key Documents

225029

Sigma-Aldrich

4-Phenylbenzonitrile

95%

Synonym(s):

4-Cyanobiphenyl, Biphenyl-4-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4CN
CAS Number:
Molecular Weight:
179.22
Beilstein:
1907924
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

liquid crystal

impurities

5% 4-bromobiphenyl

mp

85-87 °C (lit.)

SMILES string

N#Cc1ccc(cc1)-c2ccccc2

InChI

1S/C13H9N/c14-10-11-6-8-13(9-7-11)12-4-2-1-3-5-12/h1-9H

InChI key

BPMBNLJJRKCCRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A molecular dynamics study of the nematic phase of 4-n-pentyl-4'-cyanobiphenyl.
Picken, S. J., Van Gunsteren, W. F., Duijnen, P. T. V., & De Jeu, W. H.
Liq. Cryst., 6(3), 357-371 (1989)
Surface-enhanced Raman scattering of 4-cyanobiphenyl on gold and silver nanoparticle surfaces.
Joo, S. W., Chung, T. D., Jang, W. C., Gong, M. S., Geum, N., & Kim, K.
Langmuir, 18(23), 8813-8816 (2002)
Angle of twist between the two rings of 4-cyanobiphenyl when dissolved in liquid-crystalline solvents
aDe Luca, G
J. Chem. Soc., Faraday, 88(12), 1679-1684 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service