Skip to Content
Merck

Skip To

219452

3-Fluorostyrene

contains 4-tert-butylcatechol as inhibitor, 98%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Please contact Customer Service for Availability
1 900,00 CZK

About This Item

Linear Formula:
H2C=CHC6H4F
CAS Number:
Molecular Weight:
122.14
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12162002
EC Number:
206-504-9
MDL number:

1 900,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

98%

form

liquid

contains

4-tert-butylcatechol as inhibitor

refractive index

n20/D 1.517 (lit.)

bp

30-31 °C/4 mmHg (lit.)

density

1.025 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Fc1cccc(C=C)c1

InChI

1S/C8H7F/c1-2-7-4-3-5-8(9)6-7/h2-6H,1H2

InChI key

ZJSKEGAHBAHFON-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
290505729841730149
assay

98%

assay

98%

assay

93%

assay

95%

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

density

1.025 g/mL at 25 °C (lit.)

density

1.025 g/mL at 25 °C (lit.)

density

1.027 g/mL at 25 °C

density

1.023 g/mL at 25 °C

contains

4-tert-butylcatechol as inhibitor

contains

4-tert-butylcatechol as inhibitor

contains

MEHQ as inhibitor

contains

MEHQ as inhibitor

bp

30-31 °C/4 mmHg (lit.)

bp

29-30 °C/4 mmHg (lit.)

bp

-

bp

-

form

liquid

form

-

form

liquid

form

liquid


pictograms

FlameExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

84.2 °F - closed cup

flash_point_c

29 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters