20639
Di-tert-butyl 1-(tert-butylthio)-1,2-hydrazinedicarboxylate
≥99.0% (HPLC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H28N2O4S
CAS Number:
Molecular Weight:
320.45
Beilstein:
9559826
MDL number:
UNSPSC Code:
12352108
eCl@ss:
32110502
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥99.0% (HPLC)
mp
98-100 °C
application(s)
peptide synthesis
SMILES string
CC(C)(C)OC(=O)NN(SC(C)(C)C)C(=O)OC(C)(C)C
InChI
1S/C14H28N2O4S/c1-12(2,3)19-10(17)15-16(21-14(7,8)9)11(18)20-13(4,5)6/h1-9H3,(H,15,17)
InChI key
MOWYOPQOADUFLA-UHFFFAOYSA-N
Other Notes
Stable reagent for the preparation of asymmetric S-tert-butyl disulfide derivatives (e.g. for the introduction of S-tert-butylthio protection into cysteine peptides) in organic solvents
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
E Wünsch et al.
Hoppe-Seyler's Zeitschrift fur physiologische Chemie, 363(12), 1461-1464 (1982-12-01)
The sterically hindered tert-butyl thiol reacts smoothly with azodicarboxylic acid derivatives only upon addition of catalytic amounts of sodium alcoholate, yielding crystalline and analytically well characterized 1-(tert-butylthio)-1,2-hydrazinedicarboxylic acid derivatives. These sulfur-activated reagents were found to be stable on storage and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service