Skip to Content
Merck
All Photos(2)

Key Documents

205753

Sigma-Aldrich

Palladium on carbon

extent of labeling: 1 wt. % loading, 4-8 mesh

Synonym(s):

Pd/C

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
CZK 4,355.00
100 G
CZK 33,200.00

CZK 4,355.00

List PriceCZK 8,710.00Save 50%

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
CZK 4,355.00
100 G
CZK 33,200.00

About This Item

Linear Formula:
Pd
Molecular Weight:
106.42
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

CZK 4,355.00

List PriceCZK 8,710.00Save 50%

Please contact Customer Service for Availability

Request a Bulk Order

form

solid

Quality Level

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

extent of labeling

1 wt. % loading

particle size

4-8 mesh

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Stille reaction
  • Hydrogenation reactions
  • Suzuki-Miyaura cross-coupling and Heck-Mizoroki reactions
  • Deoxygenation
  • Oxidation reactions
  • Coupling reactions
Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service