Skip to Content
Merck
All Photos(1)

Documents

188212

Sigma-Aldrich

3-Methylbenzyl alcohol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH2OH
CAS Number:
Molecular Weight:
122.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.534 (lit.)

bp

215 °C/740 mmHg (lit.)

density

1.015 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(CO)c1

InChI

1S/C8H10O/c1-7-3-2-4-8(5-7)6-9/h2-5,9H,6H2,1H3

InChI key

JJCKHVUTVOPLBV-UHFFFAOYSA-N

General description

3-Methylbenzyl alcohol participates in gas phase hydrogenation of methanolic solutions of isophthaldehyde over a Ni/SiO2 catalyst.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas phase hydrogenation/hydrogenolysis of isophthaldehyde over Ni/SiO2.
Keane MA.
Bull. Soc. Chim. Belg., 104(2), 63-67 (1995)
S Fraile et al.
Journal of bacteriology, 183(19), 5571-5579 (2001-09-07)
We have isolated a recombinant phage antibody (Phab) that binds a distinct epitope of the subclass of the sigma(54)-dependent prokaryotic enhancer-binding proteins that respond directly to aromatic effectors, e.g., those that activate biodegradative operons of Pseudomonas spp. The DNA segments
Chun Chau Sze et al.
Journal of bacteriology, 184(3), 760-770 (2002-01-16)
Pseudomonas-derived regulators DmpR and XylR are structurally and mechanistically related sigma(54)-dependent activators that control transcription of genes involved in catabolism of aromatic compounds. The binding of distinct sets of aromatic effectors to these regulatory proteins results in release of a
Production of ligninolytic enzymes of the white rot fungus Panus tigrinus.
Leontievsky AA, et al.
Journal of Biotechnology, 32(3), 299-307 (1994)
Anu Vaidyanathan et al.
Journal of toxicology and environmental health. Part A, 66(12), 1133-1143 (2003-06-07)
Xylene is used as a solvent in paints, cleaning agents, and gasoline. Exposure occurs primarily by inhalation. The volatility and lipophilicity of the xylenes make the lung and nasal mucosa the primary target organs. m-Xylene (m-XYL) has been shown to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service