Skip to Content
Merck
All Photos(2)

Key Documents

184411

Sigma-Aldrich

1-Eicosene

technical grade, 80%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)17CH3
CAS Number:
Molecular Weight:
280.53
Beilstein:
1768357
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

80%

form

solid

bp

151 °C/1.5 mmHg (lit.)

mp

26-30 °C (lit.)

solubility

chloroform: soluble 5%, clear, colorless

functional group

allyl

SMILES string

CCCCCCCCCCCCCCCCCCC=C

InChI

1S/C20H40/c1-3-5-7-9-11-13-15-17-19-20-18-16-14-12-10-8-6-4-2/h3H,1,4-20H2,2H3

InChI key

VAMFXQBUQXONLZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Copolymers of 1-eicosene and ethene has been prepared using the methylaluminoxane activated dimethylsilanediyl(tetracyclopentadienyl)(tert-butylamido)-titanium dichloride catalyst system[1]. It polymerizes in the presence of zirconocene and hafnocene catalyst to yield poly-1-eicosene[2].

Application

1-Eicosene was used in the synthesis of (Z)-tetracos-5-enoic acid and racemic cis-4-(2-octadecylcyclopropane-1-yl)-butanoic acid[3].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly-α-olefins from polypropene to poly-1-eicosene made with metallocene catalysts.
Henschke O, et al.
Journal of Macromolecular Science, Part A: Pure and Applied Chemistry, 35(3), 473-481 (1998)
Geoffrey D Coxon et al.
Chemistry and physics of lipids, 126(1), 49-53 (2003-10-29)
(Z)-tetracos-5-enoic acid and racemic cis-4-(2-octadecylcyclopropane-1-yl)-butanoic acid have been prepared from 1-eicosene by a new facile route. Periodic acid cleavage of the epoxide of 1-eicosene gave nonadecanal which was condensed with 4-carboxybutyltriphenylphosphonium bromide to give predominately (Z)-tetracos-5-enoic acid. Simmons-Smith type cyclopropanation
Short and long chain branching of polyethene prepared by means of ethene copolymerization with 1-eicosene using MAO activated Me2Si (Me4Cp)(NtBu) TiCl2.
Walter P, et al.
Macromolecular Chemistry and Physics, 201(5), 604-612 (2000)
Morten T Nielsen et al.
Applied and environmental microbiology, 80(23), 7258-7265 (2014-09-23)
Forskolin is a promising medicinal compound belonging to a plethora of specialized plant metabolites that constitute a rich source of bioactive high-value compounds. A major obstacle for exploitation of plant metabolites is that they often are produced in small amounts

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service