Skip to Content
Merck
All Photos(1)

Key Documents

178381

Sigma-Aldrich

2′,3′,4′-Trichloroacetophenone

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2COCH3
CAS Number:
Molecular Weight:
223.48
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

59-64 °C (lit.)

SMILES string

CC(=O)c1ccc(Cl)c(Cl)c1Cl

InChI

1S/C8H5Cl3O/c1-4(12)5-2-3-6(9)8(11)7(5)10/h2-3H,1H3

InChI key

BXJZZJYNVIDEKG-UHFFFAOYSA-N

Application

2′,3′,4′-Trichloroacetophenone was used in the effective stimulation of 1,2,3,4-tetrachlorodibenzo-p-dioxin (TeCDD) dechlorination.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young-Beom Ahn et al.
Environmental toxicology and chemistry, 24(11), 2775-2784 (2006-01-10)
Halogenated coamendments enhanced dechlorination of 31 microM of spiked 1,2,3,4-tetrachlorodibenzo-p-dioxin (TeCDD) and 49 microM of spiked 1,2,3,4-tetrachlorodibenzofuran (TeCDF) in sediments from San Diego Bay (CA, USA) and Tuckerton (NJ, USA). Dechlorination of 1,2,3,4-TeCDD occurred to a greater extent under methanogenic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service