Skip to Content
Merck
All Photos(1)

Key Documents

178284

Ethyl 3-ethoxypropionate

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH2CH2CO2C2H5
CAS Number:
Molecular Weight:
146.18
EC Number:
MDL number:
UNSPSC Code:
51000000
PubChem Substance ID:

vapor density

5.03 (vs air)

Assay

≥99%

contains

50-150 ppm butylated hydroxytoluene as stabilizer

refractive index

n20/D 1.405 (lit.)

bp

166 °C (lit.)

density

0.95 g/mL at 25 °C (lit.)

SMILES string

CCOCCC(=O)OCC

InChI

1S/C7H14O3/c1-3-9-6-5-7(8)10-4-2/h3-6H2,1-2H3

InChI key

BHXIWUJLHYHGSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

136.4 °F

Flash Point(C)

58 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

P J Deisinger et al.
Xenobiotica; the fate of foreign compounds in biological systems, 20(10), 989-997 (1990-10-01)
1. The metabolism and disposition of ethyl 3-ethoxypropionate (EEP) in male Sprague-Dawley rats was studied following single oral gavage at 150 or 1500 mg/kg. The 14C-EEP was rapidly absorbed at both dose levels, and was excreted predominantly as metabolites in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service