Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
10 g | Please contact Customer Service for Availability | 2 120,00 CZK 1 590,00 CZK |
50 g | Please contact Customer Service for Availability | 5 040,00 CZK 3 780,00 CZK |
About This Item
Linear Formula:
C2H5C6H4NH2
CAS Number:
Molecular Weight:
121.18
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
209-594-8
Beilstein/REAXYS Number:
636282
MDL number:
Assay:
98%
Form:
liquid
1 590,00 CZK
List Price2 120,00 CZKSave 25%Web-Only Promotion
Please contact Customer Service for Availability
Quality Segment
assay
98%
form
liquid
refractive index
n
bp
212 °C (lit.)
mp
−8 °C (lit.)
density
0.975 g/mL at 25 °C (lit.)
SMILES string
CCc1cccc(N)c1
InChI
1S/C8H11N/c1-2-7-4-3-5-8(9)6-7/h3-6H,2,9H2,1H3
InChI key
AMKPQMFZCBTTAT-UHFFFAOYSA-N
1 of 1
This Item | |||
|---|---|---|---|
| description 98% | description 98% | description 98% | description ≥98% |
| assay 98% | assay 98% | assay 98% | assay ≥98% |
| Quality Level 100 | Quality Level 100 | Quality Level 200 | Quality Level 100 |
| density 0.975 g/mL at 25 °C (lit.) | density 0.975 g/mL at 25 °C (lit.) | density 0.983 g/mL at 25 °C (lit.) | density 0.968 g/mL at 25 °C (lit.) |
| mp −8 °C (lit.) | mp −5 °C (lit.) | mp −44 °C (lit.) | mp −33 °C (lit.) |
| bp 212 °C (lit.) | bp 216 °C (lit.) | bp 210 °C (lit.) | bp 231 °C (lit.) |
| form liquid | form liquid | form liquid | form liquid |
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
185.0 °F - closed cup
flash_point_c
85 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



