Skip to Content
Merck
All Photos(1)

Key Documents

174556

Sigma-Aldrich

4-Methoxy-1-naphthol

≥97%

Synonym(s):

1-Hydroxy-4-methoxynaphthalene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CZK 1,596.00
25 G
CZK 6,055.00

CZK 1,596.00

List PriceCZK 2,280.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
CZK 1,596.00
25 G
CZK 6,055.00

About This Item

Linear Formula:
CH3OC10H6OH
CAS Number:
Molecular Weight:
174.20
Beilstein:
1818465
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CZK 1,596.00

List PriceCZK 2,280.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥97%

form

solid

mp

126-129 °C (lit.)

SMILES string

COc1ccc(O)c2ccccc12

InChI

1S/C11H10O2/c1-13-11-7-6-10(12)8-4-2-3-5-9(8)11/h2-7,12H,1H3

InChI key

BOTGCZBEERTTDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methoxy-1-naphthol was used in the synthesis of 3-(4-hydroxy-1-naphthoxy)lactic acid (4-HO-NLA)[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R E Talaat et al.
Drug metabolism and disposition: the biological fate of chemicals, 14(2), 202-207 (1986-03-01)
The formation of 3-(4-hydroxy-1-naphthoxy)lactic acid (4-HO-NLA) from propranolol was investigated. Authentic 4-HO-NLA was synthesized from 4-methoxy-1-naphthol using methods previously used for preparation of naphthoxylactic acid (NLA). Cleavage of the 4-methyl ether was accomplished using iodotrimethylsilane in the presence of cyclopentene.
Denilson F Oliveira et al.
Experimental parasitology, 199, 17-23 (2019-02-23)
Exposing second-stage juveniles (J2) of Meloidogyne incognita in vitro to a phenolic compound sometimes fails to cause J2 mortality, but in tests in vivo the same compound may reduce the infectivity and population of the nematode. This work aimed to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service