Skip to Content
Merck
All Photos(1)

Key Documents

16980

Sigma-Aldrich

2-Bromododecanoic acid

≥98.0%

Synonym(s):

2-Bromolauric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CHBrCOOH
CAS Number:
Molecular Weight:
279.21
Beilstein:
1774990
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

form

solid

mp

30-32 °C

solubility

methanol: soluble 1 g/10 mL, clear, colorless

storage temp.

2-8°C

SMILES string

CCCCCCCCCCC(Br)C(O)=O

InChI

1S/C12H23BrO2/c1-2-3-4-5-6-7-8-9-10-11(13)12(14)15/h11H,2-10H2,1H3,(H,14,15)

InChI key

HXKXBCBZXXQPPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Bromododecanoic acid was used in the synthesis of carboxylic amphoteric surfactants containing ether tertiary amino and carboxylic groups. It was also used in the synthesis of C12-lipoamino acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K S Echtay et al.
Biochemistry, 40(17), 5243-5248 (2001-04-25)
The functional role of the four intrahelical arginines in uncoupling protein (UCP1) from brown adipose tissue were studied in mutants where they were replaced by noncharged residues. Wild-type and mutant UCP1 were expressed in Saccharomyces cerevisiae. As measured in isolated
S S Sidhu et al.
The Journal of physiology, 528 Pt 1, 165-176 (2000-10-06)
1. Fatty acid-induced cholecystokinin (CCK) secretion in humans and from the enteroendocrine cell line STC-1 depends critically on acyl chain length. 2. Therefore we have characterized the relationship between acyl chain length and the potency of the fatty acid to
Synthesis and properties of amphoteric surfactants.
Gawish SM, et al.
Journal of the American Oil Chemists' Society, 58(6), 757-760 (1981)
Y Pagot et al.
Applied and environmental microbiology, 62(10), 3864-3867 (1996-10-01)
This article provides information about peroxisomal fatty acid metabolism in the yeast Pichia guilliermondii. The existence of inducible mitochondrial carnitine palmitoyltransferase and peroxisomal carnitine octanoyl-transferase activities was demonstrated after culture of this yeast in a medium containing methyl oleate. The
Ying-Shan Chen et al.
Journal of pharmaceutical sciences, 102(7), 2322-2331 (2013-05-23)
Optic neuropathy is associated with retinal ganglion cell (RGC) loss leading to optic nerve damage and visual impairment. Unregulated connexin (Cx) hemichannel opening plays a role in RGC loss. Thus, inhibition via Cx43-specific mimetic peptides (MP) may prevent further cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service