Skip to Content
Merck
All Photos(1)

Key Documents

160008

Sigma-Aldrich

1-(4-Chlorophenyl)-1-cyclobutanecarbonitrile

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
CZK 5,075.00

CZK 5,075.00

List priceCZK 7,250.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
CZK 5,075.00

About This Item

Linear Formula:
ClC6H4C4H6CN
CAS Number:
Molecular Weight:
191.66
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CZK 5,075.00

List priceCZK 7,250.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.548 (lit.)

bp

295 °C (lit.)

density

1.137 g/mL at 25 °C (lit.)

functional group

chloro
nitrile

SMILES string

Clc1ccc(cc1)C2(CCC2)C#N

InChI

1S/C11H10ClN/c12-10-4-2-9(3-5-10)11(8-13)6-1-7-11/h2-5H,1,6-7H2

InChI key

XQONXPWVIZZJIL-UHFFFAOYSA-N

General description

1-(4-Chlorophenyl)-1-cyclobutanecarbonitrile is also referred as 1-(4-chlorophenyl)-1-cyanocyclobutane.[1]

Application

1-(4-Chlorophenyl)-1-cyclobutanecarbonitrile (1-(4-chlorophenyl)-1-cyanocyclobutane) can be used in the synthesis of pharmacologically active metabolites of the anti-obesity drug-sibutramine.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phase transfer alkylation of arylacetonitriles revisited.
Barbasiewicz M, et al.
Tetrahedron Letters, 47(23), 3871-3874 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service