Skip to Content
Merck
All Photos(1)

Key Documents

159719

Sigma-Aldrich

Phenylmethanesulfonyl chloride

98%

Synonym(s):

α-Toluenesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2SO2Cl
CAS Number:
Molecular Weight:
190.65
Beilstein:
972806
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
To inquire about this product 159719, please contact your local Merck office or dealer. Contact Technical Service

Assay

98%

form

solid

mp

92-94 °C (lit.)

functional group

phenyl

SMILES string

ClS(=O)(=O)Cc1ccccc1

InChI

1S/C7H7ClO2S/c8-11(9,10)6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

OAHKWDDSKCRNFE-UHFFFAOYSA-N

General description

Phenylmethanesulfonyl chloride is an aliphatic sulfonyl chloride. It reacts with triethylamine in methylene chloride or ether to yield trans-stilbene and cis diphenylethylene sulfone.[1] Its reaction with 1,1-bis(diethylamino)ethane has been studied.[2] Addition of phenylmethanesulfonyl chloride to 1,3-diphenyl-2-pyrrolidinopropene in the presence of water was studied.[3]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

THE REACTION OF PHENYLMETHANESULFONYL CHLORIDE WITH TERTIARY AMINES: FORMATION OF GEOMETRICAL ISOMERS ABOUT A CARBON-SULFUR DOUBLE BOND.
King JF and Durst T.
Canadian Journal of Chemistry, 44(7), 819-828 (1966)
Snehal Nirgude et al.
Molecules (Basel, Switzerland), 25(19) (2020-10-04)
Curcumin is known for its anticancer and migrastatic activity in various cancers, including breast cancer. Newer curcumin derivatives are being explored to overcome limitations of curcumin like low bioavailability, stability, and side effects due to its higher dose. In this
Addition of Aliphatic Sulfonyl Chlorides to Enamines. Formation of Acyclic Products.
Looker JJ.
The Journal of Organic Chemistry, 31(9), 2973-2976 (1966)
The reaction of 1,1-bs(diethylamino)ethene with phenylmethanesulfonyl chloride.
W E Truce et al.
The Journal of organic chemistry, 30(1), 71-74 (1965-01-01)
Preeti Verma et al.
The FEBS journal, 284(24), 4233-4261 (2017-10-25)
The 5-aminoimidazole-4-carboxamide ribonucleotide (AICAR) transformylase/inosine monophosphate (IMP) cyclohydrolase (ATIC) catalyzes final two steps of purine nucleotide de novo biosynthetic pathway. This study reports the characterization of ATIC from Staphylococcus lugdunensis (SlugATIC). Apart from kinetic analysis and a detailed biophysical characterization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service