Skip to Content
Merck
All Photos(1)

Key Documents

158828

Sigma-Aldrich

2-Mercapto-4(3H)-quinazolinone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6N2OS
CAS Number:
Molecular Weight:
178.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

mp

>300 °C (lit.)

SMILES string

SC1=Nc2ccccc2C(=O)N1

InChI

1S/C8H6N2OS/c11-7-5-3-1-2-4-6(5)9-8(12)10-7/h1-4H,(H2,9,10,11,12)

InChI key

PUPFOFVEHDNUJU-UHFFFAOYSA-N

General description

The FTIR and FT-Raman spectra of 2-mercapto-4(3H) quinazolinone has been studied.[1] Alkylation of 2-mercapto-4(3H)-quinazolinone has been reported.[2]

Application

2-Mercapto-4(3H)-quinazolinone was employed as a modifier for the carbon paste electrode, for voltammetric determination of mercury.[3] It was used in the synthesis of 4-quinazolinone derivatives, inhibitors of the DNA repair enzyme poly(ADP-ribose) polymerase.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stripping voltammetric determination of trace amounts of mercury using a carbon paste electrode modified with 2-mercapto-4 (3H)-quinazolinone.
Cai X, et al.
Microchimica Acta, 112(1-4), 135-146 (1993)
N Prabavathi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 129, 572-583 (2014-05-13)
The FTIR and FT-Raman spectra of 2-mercapto-4(3H) quinazolinone have been recorded in the region 4000-450 and 4000-100 cm(-1), respectively. The optimized geometry, frequency and intensity of the vibrational bands of 2-mercapto-4(3H) quinazolinone (2MQ) were obtained by the density functional theory
Synthesis and study of new 4-quinazolinone inhibitors of the DNA repair enzyme poly (ADP-ribose) polymerase (PARP).
Kulcsar G, et al.
ARKIVOC (Gainesville, FL, United States), 121, 131-131 (2003)
Tomás Lombardo et al.
Basic & clinical pharmacology & toxicology, 122(5), 489-500 (2017-12-06)
Collapse of the mitochondrial membrane potential (MMP) is often considered the initiation of regulated cell death (RCD). Carbonyl cyanide 3-chlorophenylhydrazone (CCCP) is an uncoupler of the electron transport chain (ETC) that facilitates the translocation of protons into the mitochondrial matrix
Monize M da Silva et al.
Dalton transactions (Cambridge, England : 2003), 48(39), 14885-14897 (2019-09-27)
Herein we discuss five ruthenium(ii) complexes with good cytotoxicity against cancer cells. These complexes are named [Ru(tzdt)(bipy)(dppb)]PF6 (1), [Ru(mmi)(bipy)(dppb)]PF6 (2), [Ru(dmp)(bipy)(dppb)]PF6 (3), [Ru(mpca)(bipy)(dppb)]PF6 (4) and [Ru(2mq)(bipy)(dppb)]PF6 (5), where tzdt = 1,3-thiazolidine-2-thione, mmi = mercapto-1-methyl-imidazole, dmp = 4,6-diamino-2-mercaptopyrimidine, mpca = 6-mercaptopyridine-3-carboxylic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service