Skip to Content
Merck
All Photos(3)

Documents

15299

Sigma-Aldrich

Boc-4-Abz-OH

≥98.0% (T)

Synonym(s):

4-(Boc-amino)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CO2CNHC6H4CO2H
CAS Number:
Molecular Weight:
237.25
Beilstein:
2115614
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

Assay

≥98.0% (T)

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

~200 °C (dec.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)Nc1ccc(cc1)C(O)=O

InChI

1S/C12H15NO4/c1-12(2,3)17-11(16)13-9-6-4-8(5-7-9)10(14)15/h4-7H,1-3H3,(H,13,16)(H,14,15)

InChI key

ZJDBQMWMDZEONW-UHFFFAOYSA-N

Other Notes

BOC-protected derivative of PABA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M H Norman et al.
Journal of medicinal chemistry, 39(5), 1172-1188 (1996-03-01)
A series of substituted (4-(4-(1,2-benzisothiazol-3-yl)-1-piperazinyl)butyl)benzamide derivatives was prepared and evaluated as potential atypical antipsychotic agents. The target compounds were readily prepared from their benzoyl chloride, benzoic acid, or isatoic anhydride precursors, and they were evaluated in vitro for their ability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service