Skip to Content
Merck
All Photos(1)

Documents

14799

Sigma-Aldrich

N,N′-Bis(2,6-dimethylphenyl)perylene-3,4,9,10-tetracarboxylic diimide

≥90%

Synonym(s):

DXP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H26N2O4
CAS Number:
Molecular Weight:
598.65
Beilstein:
4626671
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥90%

mp

>300 °C (lit.)

SMILES string

Cc1cccc(C)c1N2C(=O)c3ccc4c5ccc6C(=O)N(C(=O)c7ccc(c8ccc(C2=O)c3c48)c5c67)c9c(C)cccc9C

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Soluble fluorescent perylene dye having extremely high light fastness and high quantum yield

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rademacher, A., et al.
Chemische Berichte, 115, 2927-2927 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service