Skip to Content
Merck
All Photos(1)

Key Documents

147567

Sigma-Aldrich

2,3-Dinitrotoluene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)2
CAS Number:
Molecular Weight:
182.13
Beilstein:
2212428
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

mp

59-61 °C (lit.)

SMILES string

Cc1cccc(c1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C7H6N2O4/c1-5-3-2-4-6(8(10)11)7(5)9(12)13/h2-4H,1H3

InChI key

DYSXLQBUUOPLBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,3-Dinitrotoluene was used as internal standard in solid-phase microextraction method for the analysis of nitroaromatic explosives in water samples. It was used in drop assay to detect chemotactic responses toward nitroaromatic compounds for Burkholderia cepacia R34, Burkholderia sp. strain DNT and 2,4-dinitrotoluene dioxygenase mutant strain.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thammajun Leungsakul et al.
Applied microbiology and biotechnology, 69(3), 321-325 (2005-04-28)
Nitroaromatic compounds are toxic and potential carcinogens. In this study, a drop assay was used to detect chemotaxis toward nitroaromatic compounds for wild-type Burkholderia cepacia R34, wild-type Burkholderia sp. strain DNT, and a 2,4-dinitrotoluene (2,4-DNT) dioxygenase mutant strain (S5). The
A Hilmi et al.
Electrophoresis, 21(7), 1395-1404 (2000-05-29)
Amperometric detection at a bare gold electrode has been in-line coupled with capillary electrochromatography (CEC) for analysis of nitroaromatic and nitroamine explosives in contaminated soils and ground water. The CEC column packed with 3 microm C18 particles performed best using
S C Reader et al.
Toxicology and applied pharmacology, 106(2), 287-294 (1990-11-01)
The present study was undertaken to evaluate the in vitro effects of four isomers of a known testicular toxicant, dinitroluene (DNT). Rat Sertoli or Sertoli-germ cell cocultures were treated, after 3 days in culture, with DNT isomers (0.01 to 100
Neera Singh et al.
Journal of hazardous materials, 173(1-3), 343-348 (2009-09-15)
The sorption of organic contaminants in soil is mainly attributed to the soil organic matter (SOM) content. However, recent studies have highlighted the fact that it is not the total carbon content of the organic matter, but its chemical structure
Igor A Buryakov
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 800(1-2), 75-82 (2003-12-31)
Description of a gas chromatograph designed for express analysis of explosives (2,4-dinitrotoluene, 2,4,6-trinitrotoluene, pentaerythritol tetranitrate), chemical warfare agents (mustard gas, lewisite, sarin) and drugs (heroin, cocaine hydrochloride, crack) is given. The devices comprises a multicapillary chromatographic column and an ion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service