Skip to Content
Merck
All Photos(2)

Key Documents

144819

Sigma-Aldrich

1-(Trimethylsiloxy)cyclohexene

99%

Synonym(s):

1-Cyclohexenyloxytrimethylsilane, Cyclohexanone enol trimethylsilyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CZK 1,530.00

CZK 1,530.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
CZK 1,530.00

About This Item

Linear Formula:
C6H9OSi(CH3)3
CAS Number:
Molecular Weight:
170.32
Beilstein:
1859394
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
99%

CZK 1,530.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.447 (lit.)

bp

64-65 °C/15 mmHg (lit.)

density

0.875 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)OC1=CCCCC1

InChI

1S/C9H18OSi/c1-11(2,3)10-9-7-5-4-6-8-9/h7H,4-6,8H2,1-3H3

InChI key

SBEMOANGDSSPJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-(Trimethylsiloxy)cyclohexene undergoes Michael addition reaction with methyl vinyl ketone in the presence of dibutyltin bis(triflate)[1].

Application

1-(Trimethylsiloxy)cyclohexene was used in one-pot synthesis of Δ1,9- octalone[1]. It is intermediate to many complex compounds including natural products.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organotin triflate as practical catalyst for Michael addition of enol silyl ethers.
Sato T, et al.
Tetrahedron, 47(47), 9773-9782 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service