Skip to Content
Merck
All Photos(1)

Key Documents

143375

Sigma-Aldrich

5-Chloro-1-phenyl-1H-tetrazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5ClN4
CAS Number:
Molecular Weight:
180.59
Beilstein:
139070
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
97%

Assay

97%

form

solid

mp

120-123 °C (lit.)

solubility

dichloromethane: soluble 25 mg/mL, clear, colorless

SMILES string

Clc1nnnn1-c2ccccc2

InChI

1S/C7H5ClN4/c8-7-9-10-11-12(7)6-4-2-1-3-5-6/h1-5H

InChI key

DHELIGKVOGTMGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Chloro-1-phenyl-1H-tetrazole reacts with 2-hydroxyacetophenone to yield 1-phenyl-1H-tetrazol-5-oxy linked compound[1].

Application

5-Chloro-1-phenyl-1H-tetrazole was used as the reagent during the synthesis of new glycosyl donor possessing an anomeric O-(1-phenyltetrazol-5-yl) group[2].

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott P Webster et al.
Bioorganic & medicinal chemistry letters, 20(11), 3265-3271 (2010-05-11)
Inhibitors of 11beta-hydroxysteroid dehydrogenase (11beta-HSD1) show promise as drugs to treat metabolic disease and CNS disorders such as cognitive impairment. A series of 1,5-substituted 1H-tetrazole 11beta-HSD1 inhibitors has been discovered and chemically modified. Compounds are selective for 11beta-HSD1 over 11beta-HSD2
M Palme et al.
Bioorganic & medicinal chemistry, 2(11), 1169-1177 (1994-11-01)
A new glycosyl donor possessing an anomeric O-(1-phenyltetrazol-5-yl) group is prepared from 2,3,4,6-tetra-O-benzyl-D-glucose (2) and commercially available 5-chloro-1-phenyl-1H-tetrazole (1). The synthesis of glycosides derived from the donor and a few primary and secondary alcohols is reported.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service