Skip to Content
Merck
All Photos(1)

Key Documents

140198

Sigma-Aldrich

Formamidoxime

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC(=NOH)NH2
CAS Number:
Molecular Weight:
60.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

112-115 °C (lit.)

storage temp.

2-8°C

SMILES string

N\C=N\O

InChI

1S/CH4N2O/c2-1-3-4/h1,4H,(H2,2,3)

InChI key

IONSZLINWCGRRI-UHFFFAOYSA-N

Application

Formamidoxime may be used as inhibitor of DNA synthesis.

Biochem/physiol Actions

Formamidoxime undergoes oxidative cleavage of C==N bonds in tracheal smooth muscle cells catalyzed by liver cytochrome P450 producing NO[1]. It inhibits replicative DNA synthesis[2]. It has antitumor activity against L1210 leukemia[3].

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Yan et al.
Chemical communications (Cambridge, England), 48(63), 7829-7831 (2012-07-04)
The partial positive charge of amide protons is used to promote macrocyclization and form crown-ether analogs. Their deprotonation generates very selective pH-switchable alkaline earth ion receptors only in the presence of an appropriate substrate.
Y Jia et al.
The American journal of physiology, 275(5 Pt 1), L895-L901 (1998-11-14)
Nitric oxide (NO) is known to be synthesized from L-arginine in a reaction catalyzed by NO synthase. Liver cytochrome P-450 enzymes also catalyze the oxidative cleavage of C==N bonds of compounds containing a -C(NH2)==NOH function, producing NO in vitro. The
F R Althaus et al.
The Journal of biological chemistry, 256(24), 13079-13084 (1981-12-25)
Microsomal proteins from cultured chick embryo hepatocytes were separated by polyacrylamide gel electrophoresis and their rate constants of degradation (Kd) were estimated using double isotope techniques. The proteins were found to be heterogeneous in their turnover rates, proteins, or subunits
Weiwen Zhao et al.
Organic & biomolecular chemistry, 9(22), 7647-7651 (2011-09-29)
The formamidoxime configurational Z isomer coupled with the pyridylbiscarboxamide conformational codon were used to fold planar, curved structures. When embedded into macrocycles, this folded motif promotes dimerization through π-π stacking and hydrogen-bonding and the formation of tubules akin to molecular
K P Flora et al.
Cancer research, 38(5), 1291-1295 (1978-05-01)
A series of amidoximes was prepared and evaluated for possible antitumor activity against L1210 leukemia. Three of the most active compounds in the L1210 system, formamidoxime, acetamidoxime, and 2-aminoacetamidoxime hydrochloride, were also active against P388 leukemia. Acetamidoxime was marginally active

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service