Skip to Content
Merck
All Photos(1)

Documents

135712

Sigma-Aldrich

2-Chloro-5-nitrobenzonitrile

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)CN
CAS Number:
Molecular Weight:
182.56
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder

mp

105-107 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(Cl)c(c1)C#N

InChI

1S/C7H3ClN2O2/c8-7-2-1-6(10(11)12)3-5(7)4-9/h1-3H

InChI key

ZGILLTVEEBNDOB-UHFFFAOYSA-N

General description

2-Chloro-5-nitrobenzonitrile effectively conjugates with rat glutathione S-transferase (GST) isoenzymes and is an alternative model substrate in GST-research.

Application

2-Chloro-5-nitrobenzonitrile was used in online detection of rat glutathione S-transferase P1-specific inhibitors by high-resolution screening technology.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M van der Aar et al.
Xenobiotica; the fate of foreign compounds in biological systems, 26(2), 143-155 (1996-02-01)
1. Four different rat glutathione S-transferase (GST) isoenzymes, belonging to three different classes, were examined for their GSH conjugating capacity towards 11 2-substituted 1-chloro-4-nitrobenzene derivatives. Significant differences were found in their enzyme kinetic parameters Km, kcat and kcat/Km. 2. Substrates
Jeroen Kool et al.
Journal of biomolecular screening, 12(3), 396-405 (2007-03-24)
A high-resolution screening (HRS) technology is described, which couples 2 parallel enzyme affinity detection (EAD) systems for substrates and inhibitors of rat cytosolic glutathione-S-transferases (cGSTs) and purified human GST P1 to gradient reversed-phase high-performance liquid chromatography (HPLC). The cGSTs and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service