Skip to Content
Merck
All Photos(2)

Key Documents

132888

Sigma-Aldrich

2-Chloro-4′-fluoroacetophenone

99%

Synonym(s):

ω-Chloro-4-fluoroacetophenone, 4-Fluorophenacyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4COCH2Cl
CAS Number:
Molecular Weight:
172.58
Beilstein:
637860
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

47-50 °C (lit.)

functional group

chloro
fluoro
ketone

SMILES string

Fc1ccc(cc1)C(=O)CCl

InChI

1S/C8H6ClFO/c9-5-8(11)6-1-3-7(10)4-2-6/h1-4H,5H2

InChI key

UJZWJOQRSMOFMA-UHFFFAOYSA-N

Gene Information

human ... PTPN6(5777)

Looking for similar products? Visit Product Comparison Guide

General description

2-Chloro-4′-fluoroacetophenone on condensation with N-methyl-4-(methylthio)benzamidine yields 1-methyl-2-(4-methylthio)phenyl-4-(4-fluoro)phenylimidazole.

Application

2-Chloro-4′-fluoroacetophenone was used in the synthesis of S-(phenacyl)glutathiones and 2-[3-(4-chlorophenyl)-3-hydroxy-8-azabicyclo[3.2.1]oct-8-yl]-1-(4-fluorophenyl)ethanone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allergic contact dermatitis from 3-(alpha-methoxy) methylenebenzofuran-2(3H)-one (MBF) and alpha-chloro-4-fluoroacetophenone (CFAP) in chemical process workers.
M J Boffa et al.
Contact dermatitis, 34(6), 434-435 (1996-06-01)
2, 4, 5-Triarylimidazole inhibitors of IL-1 biosynthesis.
Gallagher TF, et al.
Bioorganic & Medicinal Chemistry Letters, 5(11), 1171-1176 (1995)
Donald M N Sikazwe et al.
Bioorganic & medicinal chemistry, 17(4), 1716-1723 (2009-01-22)
Arylcycloalkylamines, such as phenyl piperidines and piperazines and their arylalkyl substituents, constitute pharmacophoric groups exemplified in several antipsychotic agents. A review of previous reports indicates that arylalkyl substituents can improve the potency and selectivity of the binding affinity at D(2)-like
Philip G Board et al.
Chemical research in toxicology, 20(1), 149-154 (2007-01-18)
S-(Phenacyl)glutathione reductase (SPG-R) plays a significant role in the biotransformation of reactive alpha-haloketones to nontoxic acetophenones. Comparison of the apparent subunit size, amino acid composition, and catalysis of the reduction of S-(phenacyl)glutathiones indicated that a previously described rat SPG-R (Kitada
Justin D Smith et al.
Nature communications, 10(1), 1837-1837 (2019-04-25)
Photocatalytic polymers offer an alternative to prevailing organometallics and nanomaterials, and they may benefit from polymer-mediated catalytic and material enhancements. MPC-1, a polymer photoredox catalyst reported herein, exhibits enhanced catalytic activity arising from charge transfer states (CTSs) between its two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service