Skip to Content
Merck
All Photos(2)

Key Documents

132322

Sigma-Aldrich

m-Tolunitrile

99%

Synonym(s):

3-Methylbenzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CN
CAS Number:
Molecular Weight:
117.15
Beilstein:
507391
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.525 (lit.)

bp

99-101 °C/20 mmHg (lit.)

mp

−23 °C (lit.)

density

0.976 g/mL at 25 °C (lit.)

functional group

nitrile

SMILES string

Cc1cccc(c1)C#N

InChI

1S/C8H7N/c1-7-3-2-4-8(5-7)6-9/h2-5H,1H3

InChI key

BOHCMQZJWOGWTA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rh (III)-photosensitized interconversion of norbornadiene and quadricyclane.
Sluggett GW, et al.
The Journal of Physical Chemistry A, 101(47), 8834-8838 (1997)
Ultrasound-promoted synthesis of nitriles from aldoximes under ambient conditions.
Jiang N and Ragauskas AJ.
Tetrahedron Letters, 51(34), 4479-4481 (2010)
José Arturo Ruiz-Santoyo et al.
The Journal of chemical physics, 144(4), 044303-044303 (2016-02-02)
Rotationally resolved fluorescence excitation spectra of the origin bands in the S1 ← S0 transition of 2-tolunitrile (2TN) and 3-tolunitrile (3TN) have been recorded in the collision-free environment of a molecular beam. Analyses of these data provide the rotational constants

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service