Skip to Content
Merck
All Photos(1)

Key Documents

126314

Sigma-Aldrich

2,3-Dimethoxybenzyl alcohol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH2OH
CAS Number:
Molecular Weight:
168.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

138-142 °C/10 mmHg (lit.)

mp

48-51 °C (lit.)

functional group

hydroxyl

SMILES string

COc1cccc(CO)c1OC

InChI

1S/C9H12O3/c1-11-8-5-3-4-7(6-10)9(8)12-2/h3-5,10H,6H2,1-2H3

InChI key

CRLBBOBKCLYCJK-UHFFFAOYSA-N

Application

2,3-Dimethoxybenzyl alcohol has been used to study the solvent isotope effect (D2O versus H2O) on photomethanolysis efficiency and on fluorescence intensity[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure?reactivity studies and catalytic effects in the photosolvolysis of methoxy-substituted benzyl alcohols.
Wan P and Chak B.
J. Chem. Soc. Perkin Trans. II, 11, 1751-1756 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service