Recommended Products
Quality Level
Assay
99%
refractive index
n20/D 1.426 (lit.)
bp
159 °C (lit.)
density
0.928 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)C1CCC1
InChI
1S/C7H12O2/c1-2-9-7(8)6-4-3-5-6/h6H,2-5H2,1H3
InChI key
SMVBADCAMQOTOV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Ethyl cyclobutanecarboxylate can be used in the synthesis of cyclobutylidenecyclopropane by a three-step sequence.
Ethyl cyclobutanecarboxylate is used in C-13 NMR studies. It is closely related to cyclobutanecarbonyl chloride which is puckered with an equatorial substituent.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Carbon-13 NMR spectra of methylated cyclobutanes and ethyl cyclobutane-carboxylates.
Org. Magn. Reson., 13(3), 193-196 (1980)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service