Skip to Content
Merck
All Photos(1)

Key Documents

109908

Sigma-Aldrich

Diethylene glycol methyl ether

99%

Synonym(s):

2-(2-Methoxyethoxy)ethanol, Methyldiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
120.15
Beilstein:
1697812
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

vapor density

4.14 (vs air)

vapor pressure

0.2 mmHg ( 20 °C)

Assay

99%

autoignition temp.

221 °C

expl. lim.

1.38-22.7 %

refractive index

n20/D 1.426 (lit.)

bp

194 °C (lit.)

mp

−70 °C (lit.)

density

1.023 g/mL at 25 °C (lit.)

SMILES string

COCCOCCO

InChI

1S/C5H12O3/c1-7-4-5-8-3-2-6/h6H,2-5H2,1H3

InChI key

SBASXUCJHJRPEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Induction of r-GTP by ethylene glycol monomethyl ether.
T Kawamoto et al.
Toxicology and industrial health, 7(5-6), 473-478 (1991-09-01)
T Kawamoto et al.
Toxicology, 62(3), 265-274 (1990-06-01)
Glycol ethers have been extensively used in industry over the past 40-50 years. Numerous studies on the toxicity of glycol ethers have been performed, however, the effects of glycol ethers on the hepatic drug metabolizing enzymes are still unknown. We
E Chapoteau et al.
Clinical chemistry, 38(9), 1654-1657 (1992-09-01)
We report a colorimetric method for determining lithium in blood serum without sample pretreatment or solvent-extraction steps. The method is based on a novel chromogenic ionophore that exhibits exceptionally high selectivity (much greater than 4000:1) for lithium over sodium. The
B D Hardin et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 6(3), 430-439 (1986-04-01)
Diethylene glycol monomethyl ether (diEGME) was one of 15 glycols tested in CD-1 mice using a short-term in vivo reproductive toxicity assay (Chernoff/Kavlock test). Because results were strongly suggestive of potential reproductive toxicity, a teratology study was conducted in Sprague-Dawley
Hao Zhu et al.
Chemical communications (Cambridge, England), 48(96), 11766-11768 (2012-11-02)
Rlyso, a highly selective and sensitive pH sensor, can stain lysosomes with a novel lysosome-locating group, methylcarbitol. Rlyso was successfully used to detect lysosomal pH changes during apoptosis or induced by chloroquine while avoiding the "alkalizing effect" on lysosomes of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service