Skip to Content
Merck
All Photos(1)

Key Documents

108529

Sigma-Aldrich

Diphenylphosphinic acid

99%

Sign Into View Organizational & Contract Pricing

Select a Size

10MG
$30.70

$30.70


Usually ships in 2 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
10MG
$30.70

About This Item

Linear Formula:
(C6H5)2P(O)OH
CAS Number:
Molecular Weight:
218.19
Beilstein:
2804567
EC Number:
MDL number:
UNSPSC Code:
12352103

$30.70


Usually ships in 2 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Assay

99%

mp

193-195 °C (lit.)

SMILES string

OP(=O)(c1ccccc1)c2ccccc2

InChI

1S/C12H11O2P/c13-15(14,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H,(H,13,14)

InChI key

BEQVQKJCLJBTKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service