Přejít k obsahu
57651
Upchurch PEEK LiteTouch® Fitting
natural PEEK nut, pkg of 10 ea
Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
O této položce
UNSPSC Code:
41115700
NACRES:
NB.21
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
material
natural PEEK nut
description
litetouch fitting
packaging
pkg of 10 ea
fitting
10-32 male Upchurch nut for 1/32 in. or 1/16 in. O.D. tubing
General description
Unique design prevents twisting of PEEK tubing. The fitting consists of a special PEEK ferrule, a stainless steel ring, and a PEEK or stainless steel nut for 1/16 in. tubing. The fitting will not leak at pressures to 5000 psi/340 bar with the PEEK nut, 2500 psi/175 bar with the stainless steel nut finger-tightened or 7000 psi/480 bar with wrench tightening. After wrench-tightening the stainless steel nut, subsequent finger-tightening will hold to 5000 psi. Compatible with all manufacturers′ stainless steel nuts. Order nuts and ferrules separately.
Other Notes
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
LiteTouch is a registered trademark of IDEX Health & Science, LLC
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)