Merck
Z370819
Scienceware® replicator, 96-well
polycarbonate
Synonyma:
plate replicator, 378760002, F37876-0002, cell replication cloth, colonoy replication device, colony replication aid, colony replication tool
Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
O této položce
NACRES:
NB.22
UNSPSC Code:
41122102
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
material
polycarbonate
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37876-0002
General description
Perform tedious 96 well blotting procedures with one simple motion. The 96 "open ended" pipette tips in this unique product fit all types of plates from flat, vee or round bottom plates to 0.2 mL thin walled PCR plates and tubes. The replicator fits 96 well plates of any standard configuration, and will pick up about 10 μL from each well. Then deliver the transfer to a hybridization membrane or an agar surface. Suitable for replication of rDNA libraries, colony hybridization techniques, phage typing, etc.
Application
  • Replication of recombinant DNA libraries
  • Inoculation of filters for colony hybridization
  • PCR, phage typing
Features and Benefits
  • Tip will retain up to 10ml of fluid for placement onto filter paper, plate or other receiving media
  • Made of polycarbonate; easy-to-use
  • Reusable autoclavable
Legal Information
Scienceware is a registered trademark of Bel-Art Products, a division of SP
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)