Vybrat velikost
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
5 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 2 380,00 Kč |
O této položce
2 380,00 Kč
material
polypropylene , red
feature
positions 6 × 14
manufacturer/tradename
BRAND 4341002, BRAND
technique(s)
cryopreservation: suitable
L × W × H
265 mm × 126 mm × 38 mm
diam.
13 mm
General description
Other Notes
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| feature positions 6 × 14 | feature positions 6 × 14 | feature autoclavable at 121°C (2 bar), acc to DIN EN 258, positions 6 × 14, Holds 84 x 13 mm tubes | feature positions 6 × 14 |
| manufacturer/tradename BRAND 4341002 | manufacturer/tradename BRAND 4341001 | manufacturer/tradename BRAND 4340000 | manufacturer/tradename BRAND 4341000 |
| material polypropylene , red | material blue , polypropylene | material polypropylene | material polypropylene , white |
| L × W × H 265 mm × 126 mm × 38 mm | L × W × H 265 mm × 126 mm × 38 mm | L × W × H 265 mm × 126 mm × 75 mm | L × W × H 265 mm × 126 mm × 38 mm |
| diam. 13 mm | diam. 13 mm | diam. - | diam. 13 mm |
| technique(s) cryopreservation: suitable | technique(s) cryopreservation: suitable | technique(s) - | technique(s) cryopreservation: suitable |
Still not finding the right product?
Explore all of our products under BRAND® cryovial racks, PP
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?