O této položce
NACRES:
NB.01
UNSPSC Code:
41121815
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlimaterial
ETFE
manufacturer/tradename
BRAND 704399
fitting
GL 38 bottle, GL 45 thread
General description
For all Dispensette® models as well as seripettor® and QuikSip™
Other Notes
BR704399 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
QuikSip is a trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
Explore all of our products under Bottle adapters for BRAND® Dispensers and Burettes
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?