Vybrat velikost
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
12 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 2 180,00 Kč |
O této položce
2 180,00 Kč
material
AR-Glas®
Quality Segment
description
total delivery Ex + 15 sec waiting time
feature
class AS
packaging
pack of 12 ea
manufacturer/tradename
BRAND 29702
capacity
1 mL
General description
According to DIN EN ISO 648.
Other Notes
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description capacity 1 mL, one mark | description capacity 0.5 mL, one mark | description capacity 2 mL, one mark | description capacity 10 mL, one mark |
| capacity 1 mL | capacity 0.5 mL | capacity 2 mL | capacity 10 mL |
| feature class AS | feature class AS | feature class AS | feature class AS |
| manufacturer/tradename BRAND 29702 | manufacturer/tradename BRAND 29701 | manufacturer/tradename BRAND 29703 | manufacturer/tradename BRAND 29712 |
| material AR-Glas® | material AR-Glas® | material AR-Glas® | material AR-Glas® |
| packaging pack of 12 ea | packaging pack of 12 ea | packaging pack of 12 ea | packaging pack of 6 ea |
| description total delivery Ex + 15 sec waiting time | description total delivery Ex + 15 sec waiting time | description total delivery Ex + 15 sec waiting time | description total delivery Ex + 15 sec waiting time |
Still not finding the right product?
Explore all of our products under BRAND® BLAUBRAND® bulb pipette, calibrated to deliver (TD, EX)
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?